Assay Details
Gene Symbol: Timm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011592.2 | NP_035722.2 | 11 - 12 | 1147 | 60 | |
GenBank mRNA | AK153866.1 | - | 11 - 12 | 1152 | 60 | |
BC023454.1 | - | 11 - 12 | 1123 | 60 | ||
BC043479.1 | - | 10 - 11 | 2406 | 60 | ||
BC110677.1 | - | 11 - 12 | 1145 | 4979986 | 60 | |
BC117523.1 | - | 11 - 12 | 1137 | 40105563 | 60 | |
BC117524.1 | - | 11 - 12 | 1137 | 40105561 | 60 | |
U69898.1 | - | 11 - 12 | 1137 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
