Assay Details
| Gene Symbol: Timp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044384.1 | NP_001037849.1 | 2 - 3 | 287 | 90 | |
| NM_011593.2 | NP_035723.2 | 2 - 3 | 284 | 90 | ||
| GenBank mRNA | BC008107.1 | - | 2 - 3 | 189 | 3158222 | 90 |
| BC034260.1 | - | 2 - 3 | 189 | 3158222 | 90 | |
| BC051260.1 | - | 2 - 3 | 169 | 30056565 | 90 | |
| CJ055052.1 | - | 2 - 3 | 199 | 90 | ||
| CJ063120.1 | - | 2 - 3 | 285 | 90 | ||
| M17243.1 | - | 2 - 3 | 150 | 90 | ||
| X04684.1 | - | 2 - 3 | 226 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timp1
Genomic Map