Assay Details
| Gene Symbol: Timp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011594.3 | NP_035724.2 | 1 - 2 | 445 | 61 | |
| GenBank mRNA | AK081522.1 | - | 1 - 2 | 541 | 61 | |
| AK144236.1 | - | 1 - 2 | 360 | 61 | ||
| BC049126.1 | - | 1 - 2 | 342 | 6821553 | 61 | |
| M82858.1 | - | 1 - 2 | 340 | 61 | ||
| M93954.1 | - | 1 - 2 | 132 | 61 | ||
| X62622.1 | - | 1 - 2 | 350 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timp2
Genomic Map