Assay Details
| Gene Symbol: Timp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011594.3 | NP_035724.2 | 3 - 4 | 660 | 68 | |
| GenBank mRNA | AK032818.1 | - | 3 - 4 | 574 | 68 | |
| AK081522.1 | - | 3 - 4 | 756 | 68 | ||
| AK144236.1 | - | 3 - 4 | 574 | 68 | ||
| AY622854.1 | - | 2 - 3 | 190 | 68 | ||
| BC049126.1 | - | 3 - 4 | 557 | 6821553 | 68 | |
| M82858.1 | - | 3 - 4 | 555 | 68 | ||
| M93954.1 | - | 3 - 4 | 347 | 68 | ||
| X62622.1 | - | 3 - 4 | 565 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timp2
Genomic Map