Assay Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011596.5 | NP_035726.2 | 548 | 72 | ||
| GenBank mRNA | AB022323.1 | - | 514 | 72 | ||
| AF218252.1 | - | 533 | 72 | |||
| AK155055.1 | - | 525 | 72 | |||
| AK170122.1 | - | 535 | 72 | |||
| BC018369.1 | - | 501 | 72 | |||
| BC108992.1 | - | 445 | 40047833 | 72 | ||
| BC112905.1 | - | 508 | 3670722 | 72 | ||
| M31226.1 | - | 444 | 72 | |||
| X55184.1 | - | 445 | 72 | |||
More Information
Set Membership: |
|

Atp6v0a2
Genomic Map