Assay Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011596.5 | NP_035726.2 | 1447 | 80 | ||
| GenBank mRNA | AB022323.1 | - | 1413 | 80 | ||
| AF218252.1 | - | 1432 | 80 | |||
| AK155055.1 | - | 1424 | 80 | |||
| AK195507.1 | - | 2 - 3 | 215 | 80 | ||
| AK195863.1 | - | 2 - 3 | 216 | 80 | ||
| BC013056.1 | - | 2 - 3 | 179 | 80 | ||
| BC018369.1 | - | 1400 | 80 | |||
| BC108991.1 | - | 1344 | 40047832 | 80 | ||
| BC108992.1 | - | 1344 | 40047833 | 80 | ||
| BC112905.1 | - | 1407 | 3670722 | 80 | ||
| M31226.1 | - | 1343 | 80 | |||
| X55184.1 | - | 1344 | 80 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a2
Genomic Map