Assay Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011596.5 | NP_035726.2 | 2059 | 62 | ||
| GenBank mRNA | AB022323.1 | - | 2025 | 62 | ||
| AF218252.1 | - | 2044 | 62 | |||
| AF388674.1 | - | 2 - 3 | 233 | 62 | ||
| AK017467.1 | - | 1 - 2 | 83 | 62 | ||
| AK032909.1 | - | 1 - 2 | 813 | 62 | ||
| AK155055.1 | - | 2036 | 62 | |||
| AK170122.1 | - | 2556 | 62 | |||
| BC018369.1 | - | 2012 | 62 | |||
| BC108991.1 | - | 1956 | 40047832 | 62 | ||
| BC108992.1 | - | 1956 | 40047833 | 62 | ||
| BC112905.1 | - | 2019 | 3670722 | 62 | ||
| M31226.1 | - | 1955 | 62 | |||
| X55184.1 | - | 1956 | 62 | |||
More Information
Set Membership: |
|

Atp6v0a2
Genomic Map