Assay Details
| Gene Symbol: Atp6v0a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011596.5 | NP_035726.2 | 2591 | 66 | ||
| GenBank mRNA | AB022323.1 | - | 2557 | 66 | ||
| AF218252.1 | - | 2576 | 66 | |||
| AF388674.1 | - | 6 - 7 | 765 | 66 | ||
| AK017467.1 | - | 5 - 6 | 615 | 66 | ||
| AK032909.1 | - | 5 - 6 | 1345 | 66 | ||
| AK155055.1 | - | 2568 | 66 | |||
| AK170122.1 | - | 3088 | 66 | |||
| BC018369.1 | - | 2544 | 66 | |||
| BC108991.1 | - | 2488 | 40047832 | 66 | ||
| BC108992.1 | - | 2488 | 40047833 | 66 | ||
| BC112905.1 | - | 2551 | 3670722 | 66 | ||
| M31226.1 | - | 2484 | 66 | |||
| X55184.1 | - | 2485 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0a2
Genomic Map