Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800014777.mm.1 |
NM_001198998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014777.mm.1 |
NM_001198998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014777.mm.1 |
NM_011696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014777.mm.1 |
NM_011696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014777.mm.1 |
TC0800001871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014774.mm.1 |
NM_001198998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014774.mm.1 |
NM_001198998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014774.mm.1 |
NM_011696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014774.mm.1 |
NM_011696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014774.mm.1 |
TC0800001871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014770.mm.1 |
NM_001198998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014770.mm.1 |
NM_001198998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014770.mm.1 |
NM_011696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014770.mm.1 |
NM_011696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014770.mm.1 |
TC0800001871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014773.mm.1 |
NM_001198998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014773.mm.1 |
NM_001198998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014773.mm.1 |
NM_011696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014773.mm.1 |
NM_011696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014773.mm.1 |
TC0800001871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014775.mm.1 |
NM_001198998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014775.mm.1 |
NM_001198998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014775.mm.1 |
NM_011696.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014775.mm.1 |
NM_011696.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014775.mm.1 |
TC0800001871.mm.1 |
MTA TC |