Assay Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185164.1 | NP_001172093.1 | 3 - 4 | 541 | 56 | |
| NM_011697.3 | NP_035827.1 | 3 - 4 | 541 | 56 | ||
| GenBank mRNA | AK148188.1 | - | 3 - 4 | 540 | 56 | |
| AK178982.1 | - | 1 - 2 | 172 | 56 | ||
| AK207652.1 | - | 1 - 2 | 168 | 56 | ||
| AK208173.1 | - | 1 - 2 | 172 | 56 | ||
| AK211743.1 | - | 1 - 2 | 173 | 56 | ||
| AK216634.1 | - | 1 - 2 | 172 | 56 | ||
| BC046303.1 | - | 3 - 4 | 519 | 6492277 | 56 | |
| U43836.1 | - | 3 - 4 | 493 | 56 | ||
| U43837.1 | - | 3 - 4 | 301 | 56 | ||
| U48800.1 | - | 3 - 4 | 395 | 56 | ||
| U52820.1 | - | 3 - 4 | 395 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vegfb
Genomic Map