Assay Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011697.3 | NP_035827.1 | 5 - 6 | 650 | 103 | |
| GenBank mRNA | AK148188.1 | - | 5 - 6 | 649 | 103 | |
| AK178982.1 | - | 3 - 4 | 282 | 104 | ||
| AK207652.1 | - | 3 - 4 | 278 | 104 | ||
| AK212120.1 | - | 3 - 4 | 281 | 102 | ||
| U43836.1 | - | 5 - 6 | 602 | 103 | ||
| U52820.1 | - | 5 - 6 | 504 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vegfb
Genomic Map