Assay Details
| Gene Symbol: Zscan21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044703.2 | NP_001038168.1 | 3 - 4 | 1008 | 87 | |
| NM_001044704.2 | NP_001038169.1 | 4 - 5 | 1074 | 87 | ||
| NM_001044705.2 | NP_001038170.1 | 3 - 4 | 989 | 87 | ||
| NM_011757.3 | NP_035887.2 | 4 - 5 | 1093 | 87 | ||
| GenBank mRNA | AK146776.1 | - | 4 - 5 | 1067 | 87 | |
| AK146875.1 | - | 4 - 5 | 1054 | 87 | ||
| BC010591.1 | - | 3 - 4 | 936 | 3981599 | 87 | |
| D10630.1 | - | 3 - 4 | 927 | 87 | ||
| U41671.1 | - | 4 - 5 | 1024 | 87 | ||
| X63747.1 | - | 3 - 4 | 1784 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
oogenesis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
oogenesis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24128898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24128899 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24128897 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991440 | U41671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991440 | X63747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991440 | BC010591.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991440 | D10630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991440 | NM_011757.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40286 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825569 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84123 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103256 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142567 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991440 | U41671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991440 | X63747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991440 | BC010591.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991440 | D10630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991440 | NM_011757.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P166044 | X63747.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24128891 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103256 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05762253 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01232628 | - | - |
| Whole Mouse Genome G4122A | A_51_P166044 | X63747.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117829 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05762253 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103256 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142567 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04246129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04246131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24128899 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24128897 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24128898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24129002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24128972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04406252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04845341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04938976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04663722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04246131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24128916 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526740 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416370_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420095_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420096_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449732_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526735 | - | - |
| Mouse Genome 430 2.0 Array | 1420096_at | - | - |
| Mouse Genome 430 2.0 Array | 1416370_at | - | - |
| Mouse Genome 430 2.0 Array | 1420095_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420096_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416370_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449732_at | - | - |
| Mouse Genome 430 2.0 Array | 1420095_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449732_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22697_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22697_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500015025.mm.1 | TC0500001642.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500015025.mm.1 | NM_001044703.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015025.mm.1 | NM_001044704.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015025.mm.1 | NM_001044705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015025.mm.1 | NM_011757.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015028.mm.1 | TC0500001642.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500015028.mm.1 | NM_001044703.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015028.mm.1 | NM_001044704.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015028.mm.1 | NM_001044705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015028.mm.1 | NM_011757.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015024.mm.1 | TC0500001642.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500015024.mm.1 | NM_001044703.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015024.mm.1 | NM_001044704.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015024.mm.1 | NM_001044705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500015024.mm.1 | NM_011757.3 | Refseq |
| Mouse Expression Set 430 | 1416370_at | - | - |
| Mouse Expression Set 430 | 1416370_PM_at | - | - |
| Murine Genome U74v2 Set | 96707_at | - | - |
| Murine Genome U74v2 Set | 96707_PM_at | - | - |
| Murine Genome U74v2 Set | 162492_at | - | - |
| Murine Genome U74v2 Set | 162492_PM_at | - | - |
| Murine Genome U74v2 Set | 168368_PM_f_at | - | - |
| Murine Genome U74v2 Set | 168368_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416370_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416370_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416370_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449732_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420095_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416370_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420096_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449732_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420096_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420095_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 366747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1570687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1570687 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001570687 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2418128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2418128 | - | - |
| mouse-6 v1.1 (Illumina) | scl27086.5.1_36-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1570687 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27086.5.1_36-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570687 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2418128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3087187 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3087187 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2418128 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl27086.5.1_36-S | - | - |

Zscan21
Genomic Map