Assay Details
| Gene Symbol: Gadd45g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011817.2 | NP_035947.2 | 1 - 2 | 160 | 67 | |
| GenBank mRNA | AB021884.1 | - | 1 - 2 | 157 | 67 | |
| AF055638.1 | - | 1 - 2 | 148 | 67 | ||
| AK002237.1 | - | 1 - 2 | 158 | 67 | ||
| AK007410.1 | - | 1 - 2 | 164 | 67 | ||
| BC001989.1 | - | 1 - 2 | 141 | 3493618 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gadd45g
Genomic Map