Assay Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011848.4 | NP_035978.2 | 1 - 2 | 60 | 58 | |
| GenBank mRNA | AF099066.1 | - | 1 - 2 | 29 | 58 | |
| BU058938.1 | - | 1 - 2 | 60 | 58 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF099066.1,NM_011848.4 transcripts.
Set Membership: |
|

Nek3
Genomic Map