Assay Details
| Gene Symbol: Nek3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162947.1 | NP_001156419.1 | 7 - 8 | 651 | 95 | |
| NM_011848.4 | NP_035978.2 | 8 - 9 | 709 | 95 | ||
| GenBank mRNA | AF093416.1 | - | 7 - 8 | 656 | 95 | |
| AF099066.1 | - | 8 - 9 | 678 | 95 | ||
| BC005411.1 | - | 8 - 9 | 686 | 3590866 | 95 | |
| CV560683.1 | - | 1 - 2 | 63 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nek3
Genomic Map