Assay Details
| Gene Symbol: Slc26a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011867.3 | NP_035997.1 | 6 - 7 | 982 | 82 | |
| GenBank mRNA | AF167411.1 | - | 6 - 7 | 983 | 82 | |
| BC140995.1 | - | 5 - 6 | 830 | 9055607 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a4
Genomic Map