Assay Details
| Gene Symbol: Scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011886.2 | NP_036016.2 | 1 - 2 | 171 | 56 | |
| GenBank mRNA | AF005036.1 | - | 1 - 2 | 78 | 56 | |
| AF295403.1 | - | 1 - 2 | 76 | 56 | ||
| AK135335.1 | - | 1 - 2 | 152 | 56 | ||
| AK146204.1 | - | 1 - 2 | 173 | 56 | ||
| AK160549.1 | - | 1 - 2 | 156 | 56 | ||
| AK170115.1 | - | 1 - 2 | 138 | 56 | ||
| BC002021.1 | - | 1 - 2 | 166 | 3489293 | 56 | |
| BY170046.1 | - | 1 - 2 | 171 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scamp3
Genomic Map