Assay Details
| Gene Symbol: Scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011886.2 | NP_036016.2 | 4 - 5 | 505 | 91 | |
| GenBank mRNA | AF005036.1 | - | 4 - 5 | 412 | 91 | |
| AF295403.1 | - | 4 - 5 | 410 | 91 | ||
| AK135335.1 | - | 4 - 5 | 489 | 91 | ||
| AK146204.1 | - | 4 - 5 | 507 | 91 | ||
| AK160549.1 | - | 4 - 5 | 490 | 91 | ||
| BC002021.1 | - | 4 - 5 | 503 | 3489293 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317671 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317672 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317673 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317670 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053858 | AK170115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053858 | BC002021.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053858 | AK135335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053858 | AK160549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053858 | NM_011886.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132510 | AK170115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132510 | BC002021.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132510 | AK160549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132510 | NM_011886.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132510 | AF005036.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629188 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102661 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053858 | AK170115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053858 | BC002021.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053858 | AK135335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053858 | AK160549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053858 | NM_011886.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132510 | AK170115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132510 | BC002021.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132510 | AK160549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132510 | NM_011886.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132510 | AF005036.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317669 | - | - |
| Whole Mouse Genome G4122A | A_51_P482639 | AK170115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482639 | BC002021.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482639 | AK135335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482639 | AK146204.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482639 | AK160549.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128815 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132510 | AK170115.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132510 | BC002021.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132510 | AK160549.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132510 | NM_011886.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132510 | AF005036.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02808656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02808671 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03175247 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03175218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02808657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02808650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02808652 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03175241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02808654 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317672 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317673 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317671 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02684965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03175247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03175237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03093130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03093129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02541388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03159129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03159134 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493411 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493414 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416294_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493409 | - | - |
| Mouse Genome 430 2.0 Array | 1416294_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416294_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 24045_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 24045_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300005556.mm.1 | NM_011886.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005556.mm.1 | TC0300003214.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416294_PM_at | - | - |
| Mouse Expression Set 430 | 1416294_at | - | - |
| Murine Genome U74v2 Set | 98015_at | - | - |
| Murine Genome U74v2 Set | 98015_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416294_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416294_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 364727 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2360707 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002360707 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693421 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2693421 | - | - |
| mouse-6 v1.1 (Illumina) | scl22983.9.9_77-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2360707 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22983.9.9_77-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2360707 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245139 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245139 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl22983.9.9_77-S | - | - |

Scamp3
Genomic Map