Assay Details
| Gene Symbol: Scamp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011886.2 | NP_036016.2 | 6 - 7 | 790 | 87 | |
| GenBank mRNA | AF005036.1 | - | 6 - 7 | 697 | 87 | |
| AF295403.1 | - | 6 - 7 | 695 | 87 | ||
| AK135335.1 | - | 6 - 7 | 774 | 87 | ||
| AK146204.1 | - | 6 - 7 | 792 | 87 | ||
| AK160549.1 | - | 6 - 7 | 775 | 87 | ||
| BC002021.1 | - | 6 - 7 | 788 | 3489293 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp3
Genomic Map