Assay Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256042.1 | NP_001242971.1 | 6 - 7 | 698 | 62 | |
| NM_001256044.1 | NP_001242973.1 | 5 - 6 | 589 | 62 | ||
| NM_011939.3 | NP_036069.1 | 6 - 7 | 698 | 62 | ||
| NR_045689.1 | 6 - 7 | 698 | 62 | |||
| GenBank mRNA | AB029349.1 | - | 6 - 7 | 630 | 62 | |
| AB029350.1 | - | 6 - 7 | 625 | 62 | ||
| AK148056.1 | - | 5 - 6 | 561 | 62 | ||
| BC138130.1 | - | 6 - 7 | 698 | 8861149 | 62 | |
| BC145153.1 | - | 6 - 7 | 698 | 62 | ||
| BC145154.1 | - | 6 - 7 | 698 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsf4
Genomic Map