Assay Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256042.1 | NP_001242971.1 | 11 - 12 | 1333 | 65 | |
| NM_001256044.1 | NP_001242973.1 | 10 - 11 | 1224 | 65 | ||
| NM_011939.3 | NP_036069.1 | 11 - 12 | 1243 | 65 | ||
| NR_045689.1 | 11 - 12 | 1229 | 65 | |||
| GenBank mRNA | AB029349.1 | - | 11 - 12 | 1265 | 65 | |
| AB029350.1 | - | 11 - 12 | 1170 | 65 | ||
| AF160966.1 | - | 11 - 12 | 1260 | 65 | ||
| AK148056.1 | - | 10 - 11 | 1196 | 65 | ||
| AK184314.1 | - | 2 - 3 | 150 | 65 | ||
| AK215529.1 | - | 2 - 3 | 150 | 65 | ||
| BC138130.1 | - | 11 - 12 | 1333 | 8861149 | 65 | |
| BC145153.1 | - | 11 - 12 | 1229 | 65 | ||
| BC145154.1 | - | 11 - 12 | 1334 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsf4
Genomic Map