Assay Details
| Gene Symbol: Map3k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011946.3 | NP_036076.2 | 5 - 6 | 791 | 83 | |
| GenBank mRNA | AK143367.1 | - | 5 - 6 | 788 | 83 | |
| U43186.1 | - | 5 - 6 | 689 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k2
Genomic Map