Assay Details
| Gene Symbol: Mapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038663.1 | NP_001033752.1 | 6 - 7 | 1099 | 91 | |
| NM_011949.3 | NP_036079.1 | 6 - 7 | 1099 | 91 | ||
| GenBank mRNA | AK035386.1 | - | 6 - 7 | 1073 | 91 | |
| AK048127.1 | - | 6 - 7 | 1101 | 91 | ||
| AK087925.1 | - | 6 - 7 | 1074 | 91 | ||
| AK132241.1 | - | 6 - 7 | 1087 | 91 | ||
| AK148853.1 | - | 6 - 7 | 1061 | 91 | ||
| BC006708.1 | - | 1 - 2 | 145 | 91 | ||
| BC058258.1 | - | 6 - 7 | 1042 | 6468233 | 91 | |
| D10939.1 | - | 6 - 7 | 914 | 91 | ||
| X58712.1 | - | 6 - 7 | 872 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk1
Genomic Map