Assay Details
| Gene Symbol: Sema4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011976.1 | NP_036106.1 | 11 - 12 | 1857 | 96 | |
| GenBank mRNA | AF134918.1 | - | 11 - 12 | 1857 | 96 | |
| BC049183.1 | - | 11 - 12 | 1850 | 6395010 | 96 | |
| BC054532.1 | - | 10 - 11 | 2091 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4g
Genomic Map