Assay Details
| Gene Symbol: Anxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013470.2 | NP_038498.2 | 8 - 9 | 761 | 72 | |
| GenBank mRNA | AJ001633.1 | - | 8 - 9 | 744 | 72 | |
| AK013850.1 | - | 6 - 7 | 514 | 72 | ||
| AK090055.1 | - | 8 - 9 | 779 | 72 | ||
| AK150922.1 | - | 8 - 9 | 762 | 72 | ||
| AK150951.1 | - | 8 - 9 | 729 | 72 | ||
| AK151771.1 | - | 8 - 9 | 721 | 72 | ||
| AK152837.1 | - | 8 - 9 | 688 | 72 | ||
| AK152863.1 | - | 8 - 9 | 749 | 72 | ||
| AK166389.1 | - | 8 - 9 | 733 | 72 | ||
| AK168396.1 | - | 8 - 9 | 657 | 72 | ||
| AK168507.1 | - | 8 - 9 | 689 | 72 | ||
| AK169113.1 | - | 8 - 9 | 72 | |||
| BC090634.1 | - | 8 - 9 | 755 | 30942562 | 72 | |
| BC141167.1 | - | 8 - 9 | 715 | 9055779 | 72 | |
| CT010280.1 | - | 6 - 7 | 544 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa3
Genomic Map