Assay Details
| Gene Symbol: Anxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013470.2 | NP_038498.2 | 9 - 10 | 844 | 82 | |
| GenBank mRNA | AJ001633.1 | - | 9 - 10 | 827 | 82 | |
| AK013850.1 | - | 7 - 8 | 597 | 82 | ||
| AK090055.1 | - | 9 - 10 | 862 | 82 | ||
| AK150922.1 | - | 9 - 10 | 845 | 82 | ||
| AK150951.1 | - | 9 - 10 | 812 | 82 | ||
| AK151771.1 | - | 9 - 10 | 804 | 82 | ||
| AK152837.1 | - | 9 - 10 | 771 | 82 | ||
| AK152863.1 | - | 9 - 10 | 832 | 82 | ||
| AK166389.1 | - | 9 - 10 | 816 | 82 | ||
| AK168396.1 | - | 9 - 10 | 740 | 82 | ||
| AK168507.1 | - | 9 - 10 | 772 | 82 | ||
| AK169113.1 | - | 9 - 10 | 720 | 82 | ||
| BC090634.1 | - | 9 - 10 | 838 | 30942562 | 82 | |
| BC141167.1 | - | 9 - 10 | 798 | 9055779 | 82 | |
| CT010280.1 | - | 7 - 8 | 627 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa3
Genomic Map