Assay Details
| Gene Symbol: Atp6v0d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013477.3 | NP_038505.2 | 4 - 5 | 688 | 95 | |
| GenBank mRNA | AB088408.1 | - | 4 - 5 | 577 | 95 | |
| AK083372.1 | - | 4 - 5 | 689 | 95 | ||
| AK133921.1 | - | 4 - 5 | 696 | 95 | ||
| AK171515.1 | - | 4 - 5 | 658 | 95 | ||
| AK213362.1 | - | 1 - 2 | 79 | 95 | ||
| BC011075.1 | - | 4 - 5 | 579 | 3662404 | 95 | |
| U13840.1 | - | 4 - 5 | 577 | 95 | ||
| U21549.1 | - | 4 - 5 | 560 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v0d1
Genomic Map