Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013484.2 | NP_038512.2 | 3 - 4 | 565 | 68 | |
| GenBank mRNA | AK146812.1 | - | 3 - 4 | 566 | 68 | |
| AK201085.1 | - | 1 - 2 | 61 | 68 | ||
| BC011086.1 | - | 3 - 4 | 528 | 3992881 | 68 | |
| M57891.1 | - | 3 - 4 | 512 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C2
Genomic Map