Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013484.2 | NP_038512.2 | 15 - 16 | 2034 | 67 | |
| GenBank mRNA | AK146812.1 | - | 15 - 16 | 2035 | 67 | |
| AK206586.1 | - | 1 - 2 | 84 | 67 | ||
| AK207402.1 | - | 2 - 3 | 126 | 67 | ||
| AK213705.1 | - | 1 - 2 | 56 | 67 | ||
| BC011086.1 | - | 15 - 16 | 1997 | 3992881 | 67 | |
| M57891.1 | - | 15 - 16 | 1981 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C2
Genomic Map