Assay Details
| Gene Symbol: Cd3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013487.3 | NP_038515.3 | 3 - 4 | 507 | 105 | |
| GenBank mRNA | AK042172.1 | - | 3 - 4 | 509 | 105 | |
| AK050629.1 | - | 3 - 4 | 496 | 105 | ||
| AK153843.1 | - | 3 - 4 | 472 | 105 | ||
| AK154064.1 | - | 3 - 4 | 469 | 105 | ||
| BC062807.1 | - | 3 - 4 | 412 | 1366111 | 105 | |
| HM536173.1 | - | 3 - 4 | 407 | 105 | ||
| X02339.1 | - | 3 - 4 | 375 | 105 | ||
More Information
Set Membership: |
|

Cd3d
Genomic Map