Assay Details
| Gene Symbol: Chka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271496.1 | NP_001258425.1 | 8 - 9 | 1310 | 135 | |
| NM_013490.4 | NP_038518.2 | 7 - 8 | 1256 | 135 | ||
| NR_073190.1 | 9 - 10 | 1370 | 135 | |||
| GenBank mRNA | AB011002.1 | - | 7 - 8 | 1115 | 135 | |
| AK166465.1 | - | 7 - 8 | 1197 | 135 | ||
| BC004719.1 | - | 5 - 6 | 446 | 135 | ||
| BC060218.1 | - | 7 - 8 | 1219 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chka
Genomic Map