Assay Details
| Gene Symbol: Crabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013496.3 | NP_038524.1 | 2 - 3 | 402 | 84 | |
| GenBank mRNA | AK045283.1 | - | 2 - 3 | 344 | 84 | |
| AK144397.1 | - | 2 - 3 | 354 | 84 | ||
| CD742428.1 | - | 2 - 3 | 84 | |||
| M31552.1 | - | 1 - 2 | 55 | 84 | ||
| X15481.1 | - | 2 - 3 | 328 | 84 | ||
| X15789.1 | - | 2 - 3 | 344 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crabp1
Genomic Map