| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500033598.mm.1 |
TC0500003660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500033598.mm.1 |
NM_013559.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033598.mm.1 |
NM_013559.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033597.mm.1 |
TC0500003660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500033597.mm.1 |
NM_013559.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033597.mm.1 |
NM_013559.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033601.mm.1 |
TC0500003660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500033601.mm.1 |
NM_013559.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033601.mm.1 |
NM_013559.2 |
Refseq |