Assay Details
| Gene Symbol: Hsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013559.2 | NP_038587.2 | 12 - 13 | 1958 | 67 | |
| GenBank mRNA | AK075731.1 | - | 2 - 3 | 181 | 67 | |
| AK083179.1 | - | 1819 | 67 | |||
| AK146697.1 | - | 12 - 13 | 1877 | 67 | ||
| AK165046.1 | - | 12 - 13 | 1926 | 67 | ||
| AK172913.1 | - | 6 - 7 | 855 | 67 | ||
| BC018378.1 | - | 12 - 13 | 1833 | 3257042 | 67 | |
| D67016.1 | - | 12 - 13 | 1773 | 67 | ||
| L40406.1 | - | 12 - 13 | 1861 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsph1
Genomic Map