Assay Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013566.2 | NP_038594.2 | 12 - 13 | 2032 | 59 | |
| GenBank mRNA | AK078871.1 | - | 12 - 13 | 2033 | 59 | |
| AK144773.1 | - | 12 - 13 | 2007 | 59 | ||
| AK154766.1 | - | 12 - 13 | 2033 | 59 | ||
| AK155207.1 | - | 12 - 13 | 1999 | 59 | ||
| AK155594.1 | - | 12 - 13 | 2005 | 59 | ||
| BC011184.1 | - | 12 - 13 | 2005 | 4187781 | 59 | |
| M68903.1 | - | 12 - 13 | 1980 | 59 | ||
| M95632.1 | - | 12 - 13 | 1976 | 59 | ||
| M95633.1 | - | 12 - 13 | 1976 | 59 | ||
| S44607.1 | - | 11 - 12 | 1944 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb7
Genomic Map