Assay Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244198.1 | NP_001231127.1 | 3 - 4 | 299 | 56 | |
| NM_001244200.1 | NP_001231129.1 | 4 - 5 | 511 | 56 | ||
| NM_001244201.1 | NP_001231130.1 | 3 - 4 | 299 | 56 | ||
| NM_001244202.1 | NP_001231131.1 | 3 - 4 | 855 | 56 | ||
| NM_013627.5 | NP_038655.1 | 3 - 4 | 855 | 56 | ||
| GenBank mRNA | AF457142.1 | - | 4 - 5 | 265 | 56 | |
| AJ292077.1 | - | 2 - 3 | 123 | 56 | ||
| AJ307468.1 | - | 2 - 3 | 123 | 56 | ||
| AK028059.1 | - | 3 - 4 | 300 | 56 | ||
| AK032616.1 | - | 3 - 4 | 300 | 56 | ||
| AK045805.1 | - | 3 - 4 | 856 | 56 | ||
| BC011272.1 | - | 3 - 4 | 543 | 4008490 | 56 | |
| BC036957.1 | - | 3 - 4 | 295 | 4504106 | 56 | |
| CF725212.1 | - | 3 - 4 | 199 | 56 | ||
| X63963.1 | - | 3 - 4 | 176 | 56 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF457142.1,AJ292077.1,AJ307468.1,AK028059.1,BC011272.1,BC036957.1,NM_001244198.1,NM_001244200.1,NM_001244201.1,NM_001244202.1,NM_013627.5,X63963.1 transcripts.
Set Membership: |
|

Pax6
Genomic Map