Assay Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244198.1 | NP_001231127.1 | 4 - 5 | 422 | 57 | |
| NM_001244200.1 | NP_001231129.1 | 5 - 6 | 634 | 57 | ||
| NM_013627.5 | NP_038655.1 | 4 - 5 | 978 | 57 | ||
| GenBank mRNA | AJ292077.1 | - | 3 - 4 | 239 | 57 | |
| AJ307468.1 | - | 3 - 4 | 246 | 57 | ||
| AK032616.1 | - | 4 - 5 | 423 | 57 | ||
| BC011272.1 | - | 4 - 5 | 666 | 4008490 | 57 | |
| X63963.1 | - | 4 - 5 | 299 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax6
Genomic Map