Assay Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244198.1 | NP_001231127.1 | 7 - 8 | 848 | 63 | |
| NM_001244200.1 | NP_001231129.1 | 8 - 9 | 1060 | 63 | ||
| NM_001244201.1 | NP_001231130.1 | 6 - 7 | 806 | 63 | ||
| NM_001244202.1 | NP_001231131.1 | 6 - 7 | 1362 | 63 | ||
| NM_013627.5 | NP_038655.1 | 7 - 8 | 1404 | 63 | ||
| GenBank mRNA | AF443223.1 | - | 3 - 4 | 461 | 63 | |
| AJ292077.1 | - | 6 - 7 | 665 | 63 | ||
| AJ307468.1 | - | 6 - 7 | 672 | 63 | ||
| AK028059.1 | - | 6 - 7 | 807 | 63 | ||
| AK032616.1 | - | 7 - 8 | 849 | 63 | ||
| AK045805.1 | - | 6 - 7 | 1364 | 63 | ||
| AK139054.1 | - | 392 | 63 | |||
| BC011272.1 | - | 7 - 8 | 1092 | 4008490 | 63 | |
| BC036957.1 | - | 6 - 7 | 802 | 4504106 | 63 | |
| M77842.1 | - | 2 - 3 | 391 | 63 | ||
| X63963.1 | - | 7 - 8 | 725 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax6
Genomic Map