Assay Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244198.1 | NP_001231127.1 | 10 - 11 | 1242 | 71 | |
| NM_001244200.1 | NP_001231129.1 | 11 - 12 | 1454 | 71 | ||
| NM_001244201.1 | NP_001231130.1 | 9 - 10 | 1200 | 71 | ||
| NM_001244202.1 | NP_001231131.1 | 9 - 10 | 1756 | 71 | ||
| NM_013627.5 | NP_038655.1 | 10 - 11 | 1798 | 71 | ||
| GenBank mRNA | AF443223.1 | - | 6 - 7 | 855 | 71 | |
| AF457141.1 | - | 4 - 5 | 521 | 71 | ||
| AJ292077.1 | - | 9 - 10 | 1059 | 71 | ||
| AJ307468.1 | - | 9 - 10 | 1066 | 71 | ||
| AK028059.1 | - | 9 - 10 | 1201 | 71 | ||
| AK045805.1 | - | 9 - 10 | 1758 | 71 | ||
| AK139054.1 | - | 786 | 71 | |||
| AY064175.1 | - | 5 - 6 | 665 | 71 | ||
| BC011272.1 | - | 10 - 11 | 1486 | 4008490 | 71 | |
| BC036957.1 | - | 9 - 10 | 1196 | 4504106 | 71 | |
| BC069912.1 | - | 6 - 7 | 1233 | 71 | ||
| M77842.1 | - | 5 - 6 | 785 | 71 | ||
| X63963.1 | - | 10 - 11 | 1119 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax6
Genomic Map