Assay Details
| Gene Symbol: Prm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013638.2 | NP_038666.2 | 1 | 275 | 80 | |
| GenBank mRNA | AK132894.1 | - | 1 | 276 | 80 | |
| BC049598.1 | - | 266 | 80 | |||
| BC061127.1 | - | 274 | 6771130 | 80 | ||
| BC090986.1 | - | 335 | 6740459 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115435 | BC061127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115435 | BC090986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115435 | BC049598.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115435 | NM_013638.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115435 | AK132894.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115437 | BC061127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115437 | BC090986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115437 | BC049598.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115437 | NM_013638.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115437 | AK132894.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115437 | BC061127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115437 | BC090986.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115437 | BC049598.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115437 | NM_013638.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115437 | AK132894.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115435 | BC061127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115435 | BC090986.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115435 | BC049598.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115435 | NM_013638.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115435 | AK132894.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30544506 | - | - |
| Whole Mouse Genome G4122A | A_51_P507578 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30544421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30544506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10437679 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10437678 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422913_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10437677 | - | - |
| Mouse Genome 430 2.0 Array | 1422913_at | - | - |
| Mouse Genome 430 2.0 Array | 1422913_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19120_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19120_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600009554.mm.1 | NM_013638.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600009554.mm.1 | NM_013638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600009554.mm.1 | TC1600001200.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422913_at | - | - |
| Mouse Expression Set 430 | 1422913_PM_at | - | - |
| Murine Genome U74v2 Set | 93242_PM_at | - | - |
| Murine Genome U74v2 Set | 93242_at | - | - |
| Murine Genome U74v2 Set | 162056_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162056_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422913_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422913_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422913_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422913_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4810577 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4810577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810577 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2646845 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2646845 | - | - |
| mouse-6 v1.1 (Illumina) | scl48766.1.17_61-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48766.1.17_61-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219477 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219477 | - | - |

Prm3
Genomic Map