Assay Details
| Gene Symbol: Sema3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013657.5 | NP_038685.3 | 3 - 4 | 457 | 109 | |
| GenBank mRNA | AK034239.1 | - | 3 - 4 | 516 | 109 | |
| AK140348.1 | - | 993 | 109 | |||
| BC066852.1 | - | 3 - 4 | 457 | 30093230 | 109 | |
| X85994.1 | - | 3 - 4 | 409 | 109 | ||
More Information
Set Membership: |
|

Sema3c
Genomic Map