Assay Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083967.1 | NP_001077436.1 | 6 - 7 | 604 | 139 | |
| NM_013685.2 | NP_038713.1 | 6 - 7 | 919 | 139 | ||
| GenBank mRNA | AK028748.1 | - | 6 - 7 | 530 | 139 | |
| AK051958.1 | - | 5 - 6 | 827 | 139 | ||
| AK081012.1 | - | 6 - 7 | 556 | 139 | ||
| AK133885.1 | - | 7 - 8 | 1331 | 139 | ||
| BC043050.1 | - | 5 - 6 | 379 | 5708370 | 139 | |
| S75870.1 | - | 6 - 7 | 491 | 139 | ||
| U16322.1 | - | 6 - 7 | 891 | 139 | ||
| X91753.1 | - | 6 - 7 | 543 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcf4
Genomic Map