Assay Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083967.1 | NP_001077436.1 | 14 - 15 | 1384 | 74 | |
| NM_013685.2 | NP_038713.1 | 14 - 15 | 1699 | 74 | ||
| GenBank mRNA | AK014343.1 | - | 7 - 8 | 768 | 74 | |
| AK081012.1 | - | 14 - 15 | 1336 | 74 | ||
| AK132140.1 | - | 7 - 8 | 725 | 74 | ||
| AK133885.1 | - | 15 - 16 | 2111 | 74 | ||
| AK151433.1 | - | 7 - 8 | 670 | 74 | ||
| BC014293.1 | - | 7 - 8 | 721 | 4014231 | 74 | |
| BC043050.1 | - | 13 - 14 | 1159 | 5708370 | 74 | |
| S75870.1 | - | 14 - 15 | 1271 | 74 | ||
| U16321.1 | - | 8 - 9 | 758 | 74 | ||
| U16322.1 | - | 15 - 16 | 1671 | 74 | ||
| X91753.1 | - | 14 - 15 | 1323 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcf4
Genomic Map