Assay Details
| Gene Symbol: Tcp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085555.1 | NP_001079024.1 | 8 - 9 | 1160 | 61 | |
| NM_013687.3 | NP_038715.2 | 8 - 9 | 1397 | 61 | ||
| GenBank mRNA | BC016610.1 | - | 8 - 9 | 1380 | 61 | |
| BC058793.1 | - | 7 - 8 | 1336 | 61 | ||
| BC089527.1 | - | 8 - 9 | 1398 | 61 | ||
| BC089528.1 | - | 8 - 9 | 1143 | 6741317 | 61 | |
| X52127.1 | - | 6 - 7 | 919 | 61 | ||
| X52128.1 | - | 8 - 9 | 1375 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120334 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062024 | AK053725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986421 | BC058793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986421 | NM_001085555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986421 | NM_013687.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316378 | AK019833.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316378 | AK047821.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316378 | AK159117.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316378 | AK169949.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316378 | BC018368.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316378 | NM_126165.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28860 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145364 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100407 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986421 | BC058793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986421 | NM_001085555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986421 | NM_013687.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062024 | AK053725.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31120358 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145364 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100407 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08749753 | - | - |
| Whole Mouse Genome G4122A | A_52_P75127 | X52128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P75127 | BC089527.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P75127 | BC089528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P75127 | X52127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P75127 | BC016610.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129623 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145364 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13186169 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13084649 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13268104 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13186170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31120340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31120342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13181086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13583882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13310224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13310223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13084649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13357133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13468108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13583888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13486928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13172276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13186170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13373432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13129124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13431011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31120308 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449407 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449413 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449408 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420730_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449401 | - | - |
| Mouse Genome 430 2.0 Array | 1420730_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420730_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21463_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21463_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700016102.mm.1 | NM_001085555.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016102.mm.1 | NM_001085555.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016102.mm.1 | NM_013687.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016102.mm.1 | NM_013687.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016102.mm.1 | TC1700001750.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700016104.mm.1 | NM_001085555.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016104.mm.1 | NM_001085555.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016104.mm.1 | NM_013687.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016104.mm.1 | NM_013687.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016104.mm.1 | TC1700001750.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700016103.mm.1 | NM_001085555.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016103.mm.1 | NM_001085555.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016103.mm.1 | NM_013687.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016103.mm.1 | NM_013687.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016103.mm.1 | TC1700001750.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420730_a_at | - | - |
| Mouse Expression Set 430 | 1420730_PM_a_at | - | - |
| Murine Genome U74v2 Set | 97381_PM_s_at | - | - |
| Murine Genome U74v2 Set | 97381_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420730_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420730_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420730_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420730_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 695327 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6620670 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106100064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6620670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711249 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2711249 | - | - |
| mouse-6 v1.1 (Illumina) | scl021463.1_326-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620670 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106100064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl021463.1_326-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620670 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106100064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2951580 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239084 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2951580 | - | - |

Tcp11
Genomic Map