Assay Details
| Gene Symbol: Tec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113460.2 | NP_001106931.1 | 17 - 18 | 1841 | 73 | |
| NM_001113461.2 | NP_001106932.1 | 16 - 17 | 1775 | 73 | ||
| NM_001113464.2 | NP_001106935.1 | 16 - 17 | 1754 | 73 | ||
| NM_013689.5 | NP_038717.2 | 16 - 17 | 1734 | 73 | ||
| GenBank mRNA | AK139329.1 | - | 16 - 17 | 1750 | 73 | |
| AK154457.1 | - | 17 - 18 | 1837 | 73 | ||
| AK168452.1 | - | 16 - 17 | 1772 | 73 | ||
| AK168460.1 | - | 16 - 17 | 1771 | 73 | ||
| AK170588.1 | - | 16 - 17 | 1773 | 73 | ||
| BC018230.1 | - | 15 - 16 | 1737 | 73 | ||
| BC037071.1 | - | 16 - 17 | 1769 | 5029430 | 73 | |
| BC062884.1 | - | 12 - 13 | 1317 | 5697182 | 73 | |
| S53716.1 | - | 16 - 17 | 1765 | 73 | ||
| X55663.1 | - | 16 - 17 | 1769 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tec
Genomic Map