Assay Details
| Gene Symbol: Txk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122754.2 | NP_001116226.1 | 4 - 5 | 459 | 110 | |
| NM_001289495.1 | NP_001276424.1 | 4 - 5 | 476 | 110 | ||
| NM_013698.3 | NP_038726.2 | 3 - 4 | 543 | 110 | ||
| GenBank mRNA | AK138069.1 | - | 4 - 5 | 456 | 110 | |
| AK138786.1 | - | 4 - 5 | 473 | 110 | ||
| BC120552.1 | - | 3 - 4 | 446 | 40129475 | 110 | |
| BC137710.1 | - | 3 - 4 | 448 | 8860729 | 110 | |
| D43963.1 | - | 3 - 4 | 543 | 110 | ||
| D83201.1 | - | 3 - 4 | 483 | 110 | ||
| D83202.1 | - | 3 - 4 | 646 | 110 | ||
| U16145.1 | - | 4 - 5 | 424 | 110 | ||
| U19607.1 | - | 4 - 5 | 459 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txk
Genomic Map