Assay Details
| Gene Symbol: Txk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122754.2 | NP_001116226.1 | 10 - 11 | 1038 | 96 | |
| NM_001289494.1 | NP_001276423.1 | 9 - 10 | 972 | 96 | ||
| NM_001289495.1 | NP_001276424.1 | 10 - 11 | 1055 | 96 | ||
| NM_013698.3 | NP_038726.2 | 9 - 10 | 1122 | 96 | ||
| GenBank mRNA | AK138069.1 | - | 10 - 11 | 1035 | 96 | |
| AK138786.1 | - | 10 - 11 | 1052 | 96 | ||
| BC120552.1 | - | 9 - 10 | 1025 | 40129475 | 96 | |
| BC137710.1 | - | 9 - 10 | 1027 | 8860729 | 96 | |
| D43963.1 | - | 9 - 10 | 1122 | 96 | ||
| D83201.1 | - | 9 - 10 | 1062 | 96 | ||
| D83202.1 | - | 9 - 10 | 1225 | 96 | ||
| U16145.1 | - | 10 - 11 | 1003 | 96 | ||
| U19607.1 | - | 10 - 11 | 1038 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Txk
Genomic Map