Assay Details
| Gene Symbol: Csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289898.1 | NP_001276827.1 | 5 - 6 | 706 | 71 | |
| NM_001289899.1 | NP_001276828.1 | 5 - 6 | 1178 | 71 | ||
| NM_013767.6 | NP_038795.3 | 5 - 6 | 710 | 71 | ||
| GenBank mRNA | AB028241.1 | - | 5 - 6 | 656 | 71 | |
| AB028736.2 | - | 5 - 6 | 656 | 71 | ||
| AK158709.1 | - | 5 - 6 | 1179 | 71 | ||
| AK201751.1 | - | 1 - 2 | 127 | 71 | ||
| BC026127.1 | - | 5 - 6 | 659 | 4010696 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1e
Genomic Map