Assay Details
| Gene Symbol: Prpf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013830.2 | NP_038858.2 | 8 - 9 | 2188 | 98 | |
| GenBank mRNA | AB516304.1 | - | 8 - 9 | 2040 | 98 | |
| AF033663.1 | - | 7 - 8 | 1478 | 98 | ||
| AF283466.1 | - | 8 - 9 | 2186 | 98 | ||
| AK083926.1 | - | 8 - 9 | 2072 | 98 | ||
| BC003769.1 | - | 5 - 6 | 565 | 98 | ||
| BC042521.1 | - | 9 - 10 | 1470 | 98 | ||
| BC080864.1 | - | 8 - 9 | 2159 | 98 | ||
| BC141272.1 | - | 8 - 9 | 2114 | 9055884 | 98 | |
| BC141273.1 | - | 8 - 9 | 2114 | 9055885 | 98 | |
| U48737.1 | - | 6 - 7 | 745 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf4b
Genomic Map