Assay Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013871.3 | NP_038899.1 | 4 - 5 | 662 | 95 | |
| GenBank mRNA | AK011286.1 | - | 3 - 4 | 702 | 95 | |
| AK188121.1 | - | 2 - 3 | 95 | |||
| BC021640.1 | - | 4 - 5 | 580 | 3979667 | 95 | |
| Y13439.1 | - | 4 - 5 | 598 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk12
Genomic Map